Tetrahydrofuran-D8

Leverandør: TCI
Danger

T3502-10ML
TCIAT3502-10MLEA 6550 NOK
TCIAT3502-10ML
Tetrahydrofuran-D8
Tetrahydrofuran-D8
Formel: C₄D₈O
Kokepunkt: 65°C
Smeltepunkt: -106°C
Flash punkt: -17°C
MDL nummer: MFCD00044238
CAS-nummer: 1693-74-9
EINECS: 216-898-4
FN: 2056
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81f845ae68df75c4884471b7ffc6aab628ca09ce&vid=81f845ae68df75c4884471b7ffc6aab628ca09ce&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %
Degree of deuteration min. 99.5 atom%D


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR