Tyrphostin RG 14620 98,0 HPLC_ASSAY_METHOD

Leverandør: TCI

Synonymer: 3-(3,5-Dichlorophenyl)-2-(pyridin-3-yl)acrylonitrile

T3576-25MG T3576-100MG
TCIAT3576-25MGEA 2050 NOK
TCIAT3576-25MG TCIAT3576-100MG
Tyrphostin RG 14620 98,0 HPLC_ASSAY_METHOD
Tyrphostin RG 14620
Formel: C₁₄H₈Cl₂N₂
MW: 275.132 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00236445
CAS-nummer: 136831-49-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d69bf9eed259d52591a1be0f5f0ae88031c1d56b&vid=d69bf9eed259d52591a1be0f5f0ae88031c1d56b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 9.5 to 10.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR