4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane ≥95.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: (1-Naphthylmethyl)boronic acid pinacol ester

T3626-1G
TCIAT3626-1GEA 5125 NOK
TCIAT3626-1G
4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane ≥95.0% (by titrimetric analysis)
4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane
Formel: C₁₇H₂₁BO₂
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD22419278
CAS-nummer: 475250-57-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eae167a4f5f63d5d33e63f173a6a70895b4d0b8a&vid=eae167a4f5f63d5d33e63f173a6a70895b4d0b8a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 93.0 %
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR