3,3',5,5'-Tetrametylbenzidin løsning for ELISA, ready-to-use

Leverandør: TCI

T3854-100ML
TCIAT3854-100MLEA 795 NOK
TCIAT3854-100ML
3,3',5,5'-Tetrametylbenzidin løsning for ELISA, ready-to-use
3,3',5,5'-Tetrametylbenzidin
Formel: C₁₆H₂₀N₂
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007748
CAS-nummer: 54827-17-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorlessclear liquid
Peroxidase detection test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR