2-Undecenoic acid ≥70% (by GC)

Leverandør: TCI
Warning

U0032-5G
TCIAU0032-5GEA 1100 NOK
TCIAU0032-5G
2-Undecenoic acid ≥70% (by GC)
2-Undecenoic acid
Formel: C₁₁H₂₀O₂
MW: 184,28 g/mol
MDL nummer: MFCD00040909
CAS-nummer: 4189-02-0

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 70.0 %
Purity (Neutralization titration) min. 85.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR