Vinyl-10-undecenoate ≥92.0% (by GC) stabilised

Leverandør: TCI

Synonymer: 10-Undecenoic Acid Vinyl Ester

Stabilisering: Stabilised with 4-methoxyphenol (MEHQ)

U0057-25ML
TCIAU0057-25MLEA 915 NOK
TCIAU0057-25ML
Vinyl-10-undecenoate ≥92.0% (by GC) stabilised
Vinyl-10-undecenoate
Formel: C₁₃H₂₂O₂
MW: 210,32 g/mol
Kokepunkt: 127°C
MDL nummer: MFCD00142351
CAS-nummer: 5299-57-0

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 92.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR