N-Succinimidyl 2-(Dodecylthiocarbonothioylthio)-2-methylpropionate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: 2,5-Dioxopyrrolidin-1-yl 2-(((dodecylthio)carbonothioyl)thio)-2-methylpropanoate, 2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid N-Succinimidyl ester

U0126-1G
TCIAU0126-1GEA 1925 NOK
TCIAU0126-1G
N-Succinimidyl 2-(Dodecylthiocarbonothioylthio)-2-methylpropionate ≥98.0% (by HPLC, total nitrogen)
N-Succinimidyl 2-(Dodecylthiocarbonothioylthio)-2-methylpropionate
Formel: C₂₁H₃₅NO₄S₃
MW: 461.69 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD21363042
CAS-nummer: 925232-64-0

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR