1-Vinylimidazole bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 1-Vinylimidazolium bis(trifluoromethanesulphonyl)imide

V0145-5G
TCIAV0145-5GEA 1775 NOK
TCIAV0145-5G
1-Vinylimidazole bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
1-Vinylimidazole bis(trifluoromethanesulphonyl)imide
Formel: C₇H₇F₆N₃O₄S₂
MW: 375,26 g/mol
CAS-nummer: 1013027-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f21d958501315bf490d08424d9d282cd786cf677&vid=f21d958501315bf490d08424d9d282cd786cf677&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Yellow to Orange powder to lump to clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Melting point 35.0 to 39.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR