Transfer tanks, Transphor™

Leverandør: Cytiva
TE22
TE22EA 16875 NOK
TE22
Transfer tanks, Transphor™
Blottingapparat
Transphor™ units use transverse electrophoresis to transfer proteins from polyacrylamide gels onto nitrocellulose and PVDF membranes. The optional Transphor™ power lid delivers up to 1,5 A and 100 V in constant current mode.

  • Performs complete and even transfers of proteins
  • Transfers four 15×21 cm gels or up to sixteen 7×10 cm gels at one time
  • Cassettes load easily with even contact between gel and transfer membrane
  • Sturdy electrode panels lock safely in place automatically
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Buffervolum 1000 ml
Gelstørrelse B × L 90×100 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR