Swing Bin, Stainless Steel
Leverandør: TEKNOMEK
HWBH-SB
TEKMHWBH-SBEA 0 NOK
TEKMHWBH-SB
Swing Bin, Stainless Steel
Avfallsbøtter Avfallsbøtter, skarpe gjenstander
Our waste bin with swing lid is commonly used in hygienic environments such as healthcare centres and laboratories for sanitary waste disposal.
  • 304L-grade stainless steel
  • Brushed dull polish finish
  • Removeable swing lid top
  • Fully enclosed
Featuring a swing lid design, our own manufactured waste bin comes fully welded with 304L-grade stainless steel construction. Its strength and secure build makes it ideal for depositing waste sharps.