Wall Mounted Document Holder (A3 landscape)
Leverandør: TEKNOMEK
OFDH-A3L
TEKMOFDH-A3LEA 0 NOK
TEKMOFDH-A3L
Wall Mounted Document Holder (A3 landscape)
Dokumentholdere
Fully welded, 304-grade stainless steel A3 landscape wall mounted document holders come in a brushed dull polish finish and have a stackable design.
  • Brushed dull polish finish
  • Wall mounted recipe card/document holders
  • 1 tier stackable
  • Crevice free, easy to clean
Durable and long-lasting solution to document storage needs. Complete with drainage holes for easy cleaning, they are sturdy and hygienic.