Tall Stainless Steel Storage Cupboards
Leverandør: TEKNOMEK
SHCCTCSSDS6040 SHCCTCSDDS1040
TEKMSHCCTCSSDS6040EA 0 NOK
TEKMSHCCTCSSDS6040 TEKMSHCCTCSDDS1040
Tall Stainless Steel Storage Cupboards
Møbler Kjemikalieskap
Stainless steel cupboards 304-grade, with a range of door options ensure robustness, hygiene and ease of cleaning. Designed with hygeine first!
  • Sloping top
  • Shelves sold separately
  • 6 to 8 easy clean shelve adjustments (every 180 mm)
  • Brushed dull polish finish
  • Stainless steel adjustable feet with non-marking rubber base
Supplied with 6 (1780 mm high) easy clean shelf adjustments. The shelves have been cleverly designed so any liquid will fall to the back of the cupboard and out through the drainage holes placed in the base of the unit.
The doors areMore Product Information