Infrared thermometers, testo 835-2
Leverandør: Testo
0560.8352
TEST0560.8352EA 7200 NOK
TEST0560.8352
Infrared thermometers, testo 835-2
Termometere Digitaltermometer
Ideal for use in the glass, ceramics or the metal industry: the testo 835-T2 high temperature infrared thermometer is the ideal professional tool for a wide variety of thermal applications in the high temperature range. The testo 835-T2 has an extremely wide temperature range which allows the user to carry out thermal measurements up to 1 500 °C - and all from a safe distance. It doesn’t matter whether the objects are small, hard to reach, moving or very hot.
  • Extended temperature range for reliable measurements up to 1 500 °C
  • IR measurements from a safe