Differential pressure manometer, testo 510
Leverandør: Testo
0563.0510
TEST0563.0510EA 2325 NOK
TEST0563.0510
Differential pressure manometer, testo 510
Manometere
The pocket-sized testo 510 measures differential pressure in the range from 0 to 100 hPa. The differential pressure measurement is temperature-compensated for accurate measurement values.
  • Handy, small and easy to operate
  • 10 selectable units
  • Values can be displayed in Pascal over the entire measurement range
  • Temperature and air density compensation
  • Magnets at the rear permit free-hand work
  • Backlit display allows the measurement values to be easily read out, even in unfavourable light conditions
  • Flow velocity measurement possible with Pitot tube