Digital thermo hygrometers, testo 625
Leverandør: Testo
0563.1625
TEST0563.1625EA 3450 NOK
TEST0563.1625
Digital thermo hygrometers, testo 625
Hygrometere Digitale termohygrometere
Relative humidity and air temperature are two decisive factors for the assessment of indoor air quality and comfort. So if we want to ensure that people are comfortable indoors or that optimum conditions prevail in storage rooms, the compact testo 625 thermohygrometer is the ideal choice. To obtain precise results as quickly and easily as possible, the testo 625 not only calculates dewpoint and wet bulb temperature automatically. The time and the point averages are also displayed immediately.
  • Simple, fast and precise measurement of air temperature and relative humidity
  • Calculation of