Gas detector for ammonia
Leverandør: Testo
0563.3165
TEST0563.3165EA 8350 NOK
TEST0563.3165
Gas detector for ammonia
Lekkasjedetektorer Gassdetektorer
Constantly checks the level of ammonia for common refrigerants: When leaks are detected, the display changes from green to red.
  • Very long sensor life and permanent sensor check
  • Audible signal additionally informs of leakages detected
  • Earplug socket for secure localization of leakages in loud surroundings
Leveringsinformasjon: Set includes a sensor head, case, mains charger unit and earplug.