Lux kit, Testo 440
Leverandør: Testo
0563.4402
TEST0563.4402EA 8775 NOK
TEST0563.4402
Lux kit, Testo 440
Dataloggere
Lux kit is equipped with clearly structured measurement menu for long-term measurement and determination of illuminance according to the V-lambda curve , thus for all common light sources.
  • Designed for measuring the illuminance in workplaces
  • Turbulence measurement in accordance with EN ISO 7730 / ASHRAE 55
Non-slip probe contact surface for convenient positioning at the measuring location, internal data storage and USB port for data export. The air velocity and IAQ measuring instrument can be expanded with a large portfolio of digital probes.