Humidity kit with Bluetooth®
Leverandør: Testo
0563.4404
TEST0563.4404EA 6325 NOK
TEST0563.4404
Humidity kit with Bluetooth®
Dataloggere
Humidity kit with Bluetooth is equipped with clearly structured measurement menus for long-term measurements and parallel determination of humidity and air temperature in storage, refrigerated, and work rooms, and in ventilation ducts.
  • Convenience and maximum flexibility for duct and outlet measurements
  • Long-term monitoring of indoor air quality
  • Turbulence measurement in accordance with EN ISO 7730 / ASHRAE 55
Convenient for wireless probes, internal memory for data storage, and USB port for data export. Ideal for different range of probes forMore Product Information