delta P air flow ComboKit 1 with Bluetooth®, Testo 440
Leverandør: Testo
0563.4409
TEST0563.4409EA 21600 NOK
TEST0563.4409
delta P air flow ComboKit 1 with Bluetooth®, Testo 440
Dataloggere
Delta P air flow ComboKit 1 equipped with clearly structured measurement menu for volume flow and parallel determination of air velocity, differential pressure, humidity and temperature in ventilation ducts or at ventilation outlets.
  • Carry out measurements without any annoying cables
  • With space-saving and integrated differential pressure sensor
  • Easily transported
Convenient with wireless probes, internal data storage and USB port for data export. Ideal for long-term monitoring of indoor air quality and maximum flexibility for duct and outletMore Product Information