Cleaning Swabs for Fibre Optic Sleeve (1,25 mm)

Leverandør: Texwipe
25123X
TEXW25123XEA 6500 NOK
TEXW25123X
Cleaning Swabs for Fibre Optic Sleeve (1,25 mm)
Svabere Svabere for rengjøring
These cleaning swabs are ideal tool for cleaning 1,25 mm fiber optic sleeves, back plane connections, and polishing pucks.

  • Free of fibres, adhesives, and binders that can generate residue
  • Contains no seams or sharp surfaces that can generate particles or cause scratches
  • 1,25 mm thick head

Applications:
- Clean 1,25 mm fibre optic end faces and alignment sleeves
- Removal of slurry from 1,25 mm ferrule polishing fixtures
- Clean 1,25 mm test ports
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR