Cleaning swabs, Pillow-tip™ Coventry™
Leverandør: Texwipe
52121
TEXW52121EA 0 NOK
TEXW52121
Cleaning swabs, Pillow-tip™ Coventry™
Svabere Svabere for rengjøring
These Pillow-tip™ Coventry™ swabs consist of polyester knit fabric which is held in place using a shrink tube. They are low in nonvolatile residue, ionic and particulate contamination, while providing exceptional strength and solvent resistance.
  • Seamless design is lint-free
  • High-strength, adhesive-free swab, durable head construction
  • Excellent solvent compatibility
Engineered for high solvent retention with the cleanliness of ISO Class 4 processed polyester.
Ideal for cleaning high end disk drive components, micro mechanical cleaning, aerospace,More Product Information