Applicator swabs, Coventry™ microtips
Leverandør: Texwipe
CM502
TEXWCM502EA 0 NOK
TEXWCM502
Applicator swabs, Coventry™ microtips
Svabere Svabere for prøvetaking
Coventry™ microtips are designed for precision applications needing pinpoint accuracy. The tapered porous polyvinylidene fluoride tip is effective as a stylus or pick for areas with tight tolerances.
  • Low in extractables and low particulate levels
  • Compatible with most solvents, adhesives, coatings, and inks
  • Assembled without adhesives or glues
Used for dispensing and dispersing solvents, adhesives and primers.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904