Cleanroom wipes, pre-saturated, TechniSat® Gowning Room Wipe™
Leverandør: Texwipe
TX1048
TEXWTX1048EA 12900 NOK
TEXWTX1048
Cleanroom wipes, pre-saturated, TechniSat® Gowning Room Wipe™
Papirservietter Renromsservietter Våtservietter, renrom
This wipe made of 55% non woven cellulose and 45% polyester fibre is pre-saturated with 6% IPA.
  • Ultra low particle and extractable levels
  • Laundered cut edge
  • Delivers pre-measured amount of high-purity alcohol to the surface: Optimises alcohol usage and reduces spill potential
  • Solvent-safe cleanroom packaging
  • Each case is provided with a tub for convenient dispensing
Ideal for cleaning the exterior of production and laboratory equipment in cleanrooms and for cleaning surfaces during and following the production flow.
Suitable for use in Class 100,More Product Information