Cleaning swabs
Leverandør: Texwipe
TX805 TX803 TX804
TEXWTX805EA 840 NOK
TEXWTX805 TEXWTX803 TEXWTX804
Cleaning swabs
Svabere Svabere for rengjøring
General-purpose swabs constructed from medical grade PU foam.
  • Thermal bond construction eliminating the risk of adhesive contamination
  • Can pick up powders and dusts
  • Lot coded for traceability and quality control
Convenient use with adhesives, greases and liquids, in production.
Forpakning: In a reclosable silicone-free bag.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904.