Cleanroom wipes, pre-saturated, PolySat®
Leverandør: Texwipe
TX8723
TEXWTX8723EA 12025 NOK
TEXWTX8723
Cleanroom wipes, pre-saturated, PolySat®
Papirservietter Renromsservietter Våtservietter, renrom
These 100% melt-blown PP wipes are pre-wetted with a 0,2 µm filtered, 70% isopropyl alcohol/30% deionized water solution.
  • Softness protects sensitive surfaces
  • Designed for use in small spaces and confined work areas
  • Secure, reclosable packaging for reduced evaporation
  • Low particle generation, low levels of solvent extractables
Ideal for cleaning sensitive surfaces, parts and products in medical device manufacturing, in microelectronics, in pharmaceutical environments and in food processing areas. Also suitable for use in gowning rooms.
Suitable forMore Product Information