Cytocentrifuges, Cellspin®
Leverandør: THARMAC GmbH
JC1000-8-G-R JC1000-12-L-R JC2000-8-G-R JC1000-4-S-R JC2000-8-G-D JC1000-4-L-R JC1000-4-G-R JC1000-8-S-R JC1000-8-L-R JC1000-12-S-R JC1000-12-G-R JC1000-12-EG-R JC1000-4-S-D JC1000-4-L-D JC1000-8-G-D JC1000-12-L-D JC2100-12-L-R JC2100-12-G-R JC1000-4-G-D JC1000-8-S-D JC1000-8-L-D JC1000-12-S-D JC1000-12-G-D JC1000-12-EL-R JC1000-12-EG-D JC2000-12-EL-D JC2000-12-EG-D JC2000-12-L-R JC2000-12-G-R JC2000-12-G-D JC2100-12-L-D JC2100-12-G-D JC2000-8-L-R JC2000-8-L-D JC2100-12-EL-R JC2100-12-EL-D JC2100-12-EG-R JC2100-12-EG-D
521-1995EA 0 NOK
521-1995 720-1782 THARJC2000-8-G-R 521-1994 THARJC2000-8-G-D 521-1993 521-1991 521-1997 521-1996 521-1990 521-1989 521-1988 720-2930 521-1992 THARJC1000-8-G-D THARJC1000-12-L-D 521-2056 521-2055 THARJC1000-4-G-D THARJC1000-8-S-D THARJC1000-8-L-D THARJC1000-12-S-D THARJC1000-12-G-D THARJC1000-12-EL-R THARJC1000-12-EG-D THARJC2000-12-EL-D THARJC2000-12-EG-D THARJC2000-12-L-R THARJC2000-12-G-R THARJC2000-12-G-D THARJC2100-12-L-D THARJC2100-12-G-D THARJC2000-8-L-R THARJC2000-8-L-D THARJC2100-12-EL-R THARJC2100-12-EL-D THARJC2100-12-EG-R THARJC2100-12-EG-D
Cytocentrifuges, Cellspin®
Sentrifuger Sentrifuger, bordmodell
Versatile cytocentrifuges suitable for small and large sample volumes, designed for thin layer and mono layer cell preparations in medical applications such as cytology, urology, microbiology, haematology, immunocytochemistry, virology and oncology.
  • Quick change of removable rotors within seconds
  • Smooth running, high performance motor featuring a maintenance-free frequency drive
  • Powered lid locking, quick entry foil keypad with easy to read, digital display and twist knob to enter values
  • Cellspin II (refrigerated): Variable temperature setting from –20 to +