Automated stainers, Cellstain® 6
Leverandør: THARMAC GmbH
JC651-30
THARJC651-30EA 0 NOK
THARJC651-30
Automated stainers, Cellstain® 6
Fargetrau for objektglass
Automated stainer with six staining troughs for all staining protocols with up to six different chemicals (May-Grünwald, Giemsa, Wright, Pappenheim, Leishman, Gram staining).
  • Pre-programmed with six staining programs
  • Programs protected by a password
  • Very low consumption of stains and reagents
  • The agitation and the drain time is freely selectable
  • Linear staining machine to move the slide holder forward and backward
Suitable for haematology and bacteriology staining. Due to its compact size, it can be placed under a fume hood; a compatible fumeMore Product Information