Automated stainer, Cellstain® 15
Leverandør: THARMAC GmbH
JC665-30
THARJC665-30EA 0 NOK
THARJC665-30
Automated stainer, Cellstain® 15
Fargetrau for objektglass
The Cellstain 15 is a fully automated stainer for Papanicolaou, HE and other stainings.
  • 20 programmable procedures in the memory
  • Low transfer of reagents through the slide holder
  • Small format fitting under any existing fume hood
  • Agitation and drain time are freely selectable
  • Moveable slide holder to enable the user to program a large number of staining programs
Bestillingsinformasjon: Supplied with 15 staining troughs, slide holder and shortened trough for loading and unloading.