Automated stainer, Cellstain® 23
Leverandør: THARMAC GmbH
JC800-30
THARJC800-30EA 0 NOK
THARJC800-30
Automated stainer, Cellstain® 23
Fargetrau for objektglass
The CellStain 23 features programming unit for individually adjustable steps in staining, rinsing and drying stations and implements of different stainings due to forwards and backwards movement of slide holder. It contains storage place for 20 staining programs up to 30 steps each with password protected access.
  • Low transfer of reagents through the slide holder
  • Small format fits under any existing fume hood
  • Optional drying station with programmable temperature
Leveringsinformasjon: Supplied with 23 staining troughs (500 ml) with lid, one slide holderMore Product Information