CryoPlus™ Rack for Cryogenic Vials
Leverandør: Thermo Fisher Scientific
4000005
THBR4000005EA 3525 NOK
THBR4000005
CryoPlus™ Rack for Cryogenic Vials
Stativer for rør Kryorørstativer
CryoPlus™ series racks are ideal for use with CryoPlus™ LN2 storage systems.
  • Rack includes: cardboard boxes and cardboard dividers for 5 ml vials
  • 6 cardboard boxes with 100 cell divider
  • Holds: 6 boxes/rack