Kaliumsulfat, tablets, PS KJELTABS mineralisation catalyst

Leverandør: THOMPSON & CAPPER

AA23
THOMAA23EA 7050 NOK
THOMAA23
Kaliumsulfat, tablets, PS KJELTABS mineralisation catalyst
Potassium sulphate
Formel: K₂SO₄
MW: 174.26 g/mol
Kokepunkt: 1698
Smeltepunkt: 1069
Tetthet: 2.66 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011388
CAS-nummer: 7778-80-5
EINECS: 231-915-5
Merck Indeks: 13,07759

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR