Natriumsulfat, tablets, KJELTABS NA mineralisation catalyst

Leverandør: THOMPSON & CAPPER

Synonymer: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

AA40
THOMAA40EA 4125 NOK
THOMAA40
Natriumsulfat, tablets, KJELTABS NA mineralisation catalyst
Sodium sulphate
Formel: Na₂SO₄
MW: 142.04 g/mol
Kokepunkt: 1429
Smeltepunkt: 888
Tetthet: 2.7 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003504
CAS-nummer: 7757-82-6
EINECS: 231-820-9
Merck Indeks: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR