Cleanroom dry vacuum cleaner, CRW-10EX (4W)
Leverandør: TIGER-VAC
110540B
TIGV110540BEA 76300 NOK
TIGV110540B
Cleanroom dry vacuum cleaner, CRW-10EX (4W)
Vakuumrensere
Wet and dry recovery cleaner designed for the recovery of broken ampules and glass in contamination controlled environments. The broken ampules and glass are separated from liquid and they are recovered in an autoclavable sieve basket made of electropolished stainless steel AISI 304. The sieve basket allows for easy and safe disposal of recovered materials. This unit is cleanroom compatible and can be used in the pharmaceutical industry.
  • Single phase by-pass motor
  • Based on a 4-wheel dolly for easy maneuverability
  • Recovery tank is autoclavable
  • An optional