Vacuum cleaner, B-8 RE HEPA, Tiger-Vac®
Leverandør: TIGER-VAC
110560B
TIGV110560BEA 19775 NOK
TIGV110560B
Vacuum cleaner, B-8 RE HEPA, Tiger-Vac®
Vakuumrensere
This unit is designed for general cleanup: for asbestos, silica, lead, and other toxic/nuisance dusts recovery.
  • By-Pass heavy duty motor
  • Equipped with 4 wheels
  • Liquid recovery capacity of 30 liters
  • Silent operation
This unit contains a true HEPA Filter (H14) located downstream outside of the motor. It is equipped with an disposable filter bags for easy and safe disposal of recovered material.
Construction: Polyethylene recovery tank
NB: Note: For wet and dry recovery
Bestill nå
SPESIFIKASJONER
Modell B-8 (F) RE HEPA
Environmentally Preferable
luftflyt 283,33 l/min
Vakuumtrykk 2100 mm H2O
Hestekrefter 1,8 hp
Fase Single
Spenning 230 V
Frekvens 50 Hz
Strøm 5 A,1400 W
Strøm 5 A,1400 W
B×D×H 580×460×710
Vekt 18 kg