Vacuum cleaner, 2D-9 NUC HEPA, Tiger-Vac®
Leverandør: TIGER-VAC
110626B
TIGV110626BEA 57000 NOK
TIGV110626B
Vacuum cleaner, 2D-9 NUC HEPA, Tiger-Vac®
Vakuumrensere
This unit contains a true nuclear grade HEPA Filter (H14) constructed and tested to methods consistant with ASME AG-1. An external ULPA filter (U15) is available.
  • Disposable containment drum assemblies
  • Electrically operated
  • Removable and reusable power head
Construction: Polycarbonate recovery tank.
NB: Note: For dry recovery only.