Industrial vacuum cleaner, 2D-25L (DT) MFS WT
Leverandør: TIGER-VAC
111403B
TIGV111403BEA 30325 NOK
TIGV111403B
Industrial vacuum cleaner, 2D-25L (DT) MFS WT
Vakuumrensere
This high efficeincy vacuum cleaner has been designed for general cleanup for the food industry.
  • Bypass heavy duty motor
  • Manual Filter Shaker (MFS) system
  • Detachable Recovery Tank (DT) for easy disposal of recovered materials
  • Equipped with a 4-Wheel cart and tool basket
  • Cart has SD (static dissipating) conductive wheels
Construction: Heavy duty powder coated body and stainless steel recovery tank (SS AISI 430).
Sertifiseringer: All HEPA vacuum cleaners are aerosol leak tested before leaving the facility.
Leveringsinformasjon: Includes a completeMore Product Information