Industrial vacuum cleaner, 2D-25L (DT) MRP WT
Leverandør: TIGER-VAC
111413B
TIGV111413BEA 34650 NOK
TIGV111413B
Industrial vacuum cleaner, 2D-25L (DT) MRP WT
Vakuumrensere
This industrial vacuum cleaner is designed for the food industry.
  • Bypass heavy duty motor
  • Manual Reverse Purge (MRP) with SD (static dissipating) reverse purge cartridge
  • Detachable Recovery Tank (DT) for easy disposal of recovered materials
  • Equipped with a 4-wheel cart and tool basket
  • Cart has SD (static dissipating) conductive wheels
Construction: Heavy duty powder coated body and stainless steel (AISI 430) recovery tank.
Sertifiseringer: All HEPA vacuum cleaners are aerosol leak tested before leaving our facility.
Leveringsinformasjon: More Product Information
Bestill nå
SPESIFIKASJONER
Modell 2D-25L (DT) MRP WT
Environmentally Preferable
luftflyt 153 m3/h
Vakuumtrykk 2030 mm H2O
Filtre HEPA Filter - Included with an efficiency of 99,995% on 0,3 micron. Tested: IEST RP-CC001. H14 by MPPS method as per EN 1822 and OSHA compliant.
Gjenvinning Dry recovery-tank: 24 l
Støynivå 75 dB(A)
Lengde ledning 10,6 m
Fase Single
Spenning 230 V
Frekvens 50 Hz
Strøm 5 A,1200 W
Strøm 5 A,1200 W
B×D×H 580×530×1020 mm
Vekt 23 kg