Vacuum Cleaner, CD-230V EX (CFB) PHARMA
Leverandør: TIGER-VAC
112162B3
TIGV112162B3EA 0 NOK
TIGV112162B3
Vacuum Cleaner, CD-230V EX (CFB) PHARMA
Vakuumrensere
Designed to be attached to process machines and for general cleanup. Can also be used for the recovery of tablets in the pharmaceutical industry.
  • Legally certified explosion proof/dust ignition proof
  • Less than 10 Ω of resistivity
  • For dry recovery only
- ULPA filter with an efficiency of 99,999% at 0,12 micron.
- Filtration efficiency of 99,9995% at 0,18 micron.
- Tested IEST-RP-CC001. U15 by MPPS method as per EN 1822.
- All stainless steel type 304 construction.
Available with a conductive filter bag (CFB) or manual reverse purge with filter tubes (More Product Information
Bestill nå
SPESIFIKASJONER
Modell CD-230V EX (CFB) PHARMA
Environmentally Preferable
luftflyt 212 m³/h
Vakuumtrykk 2540 mm H20
Filtre ULPA filter with an efficiency of 99,999% at 0,12 micron
Filtration efficiency of 99,9995% at 0,18 micron
Tested IEST-RP-CC001
U15 by MPPS method as per EN 1822
Lengde ledning 10 m
Fase Single
Spenning 230 V
Frekvens 50 Hz
Strøm 1500 W,12,3 A
Strøm 1500 W,12,3 A
B×D×H 480×640×1500 mm
Vekt 97 kg