Tape dispenser
Leverandør: PRECISION DYNAMICS TIMEM
3161
TIME3161EA 1350 NOK
TIME3161
Tape dispenser
Etikett- og tapedispensere
This dispenser is suitable for a wide variety of labeling applications. It is designed to keep the work station and drawer clutter-free.
  • Made of sturdy plastic
  • Heavy duty with weighted base
  • Adjustable for rolls with 25 mm (1”) or 76 mm (3”) cores and 12,7 mm (¹/₂”), 19 mm (³/₄”) and 25 mm (1”) widths