Spectrophotometric colorimeters, model AF, Lovibond®
Leverandør: Lovibond Tintometer
107103
TINT107103EA 0 NOK
TINT107103
Spectrophotometric colorimeters, model AF, Lovibond®
Kolorimeter Kolorimeter med Automatisk Transmittans
The AF710-3 is a visual instrument used for colour grading of oils and fats.
Colour is determined by comparing light transmitted through a column of oil with that transmitted through a series of Red and Yellow glasses calibrated in accordance with the AOCS-Tintometer® Colour Scale.
The glasses enable a colour match to be found from the light passing through the oil. This colour is then expressed in units of Red and Yellow.
Bestill nå
SPESIFIKASJONER
Modell Model AF
Lyskilde 2×12 Volt, 20 W tungsten halogen lamp
Environmentally Preferable
Lystyper Approximates to daylight
Optisk system Visual colour comparison against colour glasses
B×D×H 330×100×480 mm
Vekt 8,3 kg