Absorptiometer cell
Leverandør: Lovibond Tintometer
605534
TINT605534EA 9300 NOK
TINT605534
Absorptiometer cell
Kyvetter Spesialkyvetter
Absorptiometer cell made of optical glass with path length of 10 mm.