Cation exchange chromatography columns, TSK-GEL® SP-NPR
Leverandør: TOSOH Bioscience
0013076
TOYO0013076EA 17575 NOK
TOYO0013076
Cation exchange chromatography columns, TSK-GEL® SP-NPR
Kromatografikolonner
TSK-Gel® SP-NPR are polymer based stainless steel columns. These columns provide fast separations due to their small spherical particles.
  • Nonporous columns
  • Used in gene therapy research