Cation exchange resins, Toyopearl® and TSK-GEL®
Leverandør: TOSOH Bioscience
0007474 0007475 0007991 0007994 0007997 0008437 0014028 0014714 0014716 0043202 0043203 0043272 0043382 0021805 0021834 0014705 0019803 0019822 0021835 0014699 0014695 0014696 0014697 0014700 0014698 0043282 0021804 0021806 0021946 0021947 0021948
TOYO0007474EA 13775 NOK
TOYO0007474 TOYO0007475 TOYO0007991 TOYO0007994 TOYO0007997 TOYO0008437 TOYO0014028 TOYO0014714 TOYO0014716 TOYO0043202 TOYO0043203 TOYO0043272 TOYO0043382 TOYO0021805 TOYO0021834 TOYO0014705 TOYO0019803 TOYO0019822 TOYO0021835 TOYO0014699 TOYO0014695 TOYO0014696 TOYO0014697 TOYO0014700 TOYO0014698 TOYO0043282 TOYO0021804 TOYO0021806 TOYO0021946 TOYO0021947 TOYO0021948
Cation exchange resins, Toyopearl® and TSK-GEL®
Kromatografiabsorbenter HPLC sorbenter
Cation exchange resins are polymer based media developed for the separation of biomolecules including monoclonal antibodies and peptides.
TOYOPEARL CM-650 is a weak cation exchanger for biomolecule purification. It is available in three grades and can be used for high throughput capture, intermediate purification, and polishing process steps.
TOYOPEARL® SP-650 is a strong cation exchanger for preparative purification. It can be used for initial, intermediate, final purification process.
TOYOPEARL® SP-550 is is a strong cation exchanger useful for preparative purification of insulin and proteins with molecular weights up to 100,000. It is used for initial refining process.
TSKgel SP-5PW is a strong cation exchanger for preparative purification. It is a small particle size filler for final purification process and high performance separation process.
TOYOPEARL® GigaCap CM-650 is a highly adsorptive cation exchanger for protein purification. Suitable for initial purification (capture) process and intermediate refining process, it shows a high adsorption amount for a wide range of samples such as antibodies, general proteins, peptides etc. It shows different selectivity from S (SP) type cation exchanger.