Cation exchange chromatography columns, TSK-GEL® Bioassist S
Leverandør: TOSOH Bioscience
0019686
TOYO0019686EA 22000 NOK
TOYO0019686
Cation exchange chromatography columns, TSK-GEL® Bioassist S
Kromatografikolonner
TSK-Gel® BioAssist S polymer based packed PEEK columns are designed for the separation of large biomolecules. Suitable for use in HPLC, laboratory and semi-preparative applications.
  • Very large pores of the column offers high capacity and resolution at a low column pressure drop
  • Polymerisation technique is used for homogeneous distribution of ion exchange groups
  • Novel bonded phase design provides high dynamic capacity