Affinity chromatography column for fast evaluation of ADCC activity, TSKgel® FcR-IIIA-NPR
Leverandør: TOSOH Bioscience
0023513
TOYO0023513EA 31475 NOK
TOYO0023513
Affinity chromatography column for fast evaluation of ADCC activity, TSKgel® FcR-IIIA-NPR
Kromatografikolonner
TSKgel® FcR-IIIA-NPR is based on a recombinant Fc gamma IIIa receptor ligand immobilised on a non-porous polymer particle. It allows fast assessment of biologic activity of monoclonal antibodies.
  • Easy and reproducible HPLC analysis based on FcγRIIIa affinity of mAbs
  • Unique glycoprotein elution profile of IgG allows assessment of ADCC activity
  • Applicable to purified samples and cell culture supernatant alike
  • Fast cell line screening, upstream development, lot-to-lot comparison
Fcγ IIIa receptor plays a key role in antibody-dependent cellmediatedMore Product Information