Multi-Angle Light-Scattering Detector, LenS3™
Leverandør: TOSOH Bioscience
0040000 0040001
TOYO0040000EA 0 NOK
TOYO0040000 TOYO0040001
Multi-Angle Light-Scattering Detector, LenS3™
Kromatografidetektorer
Tosoh’s LenS3™ Multi-Angle Light-Scattering detector combines high-sensitivity MW and Rg determinations with flexibility and ease of use.
  • Characterisation of molecules down to 200 to 10⁷ Da and Rg down to <2 to >300 nm
  • Optics design for low noise and high scattering intensity
  • Compatible with GPC, SEC, HPLC and UHPLC systems
  • Compatible with all common solvents (THF, HFIP, aqueous salt buffers)
  • Modular software for simultaneous acquisition data analysis, sequence editing
  • Analysis modules for step-by-step guidance
  • Proprietary conical flow path
  • Laser:
More Product Information