Hydrophobic interaction chromatography resins, Toyopearl®, TSK-GEL®
Leverandør: TOSOH Bioscience
0043150 0043175 0019806 0043125 0043278
TOYO0043150EA 9325 NOK
TOYO0043150 TOYO0043175 TOYO0019806 TOYO0043125 TOYO0043278
Hydrophobic interaction chromatography resins, Toyopearl®, TSK-GEL®
Kromatografiabsorbenter HPLC sorbenter
These are bulk media products which contain three or four different ligand types offered for a chromatography mode.
Physical properties of resins in different buffer systems can be identified by slurry and slurry mechanics, resin handling during column packing, mechanical strength relative to other resin backbones and degree of compressibility.
It is used to determine the resin's dynamic binding capacity, selectivity, column efficiency, operating conditions by developmental scientist or engineer.