Manual detergent, alkaline, Sapphire
Leverandør: TRISTEL SOLUTIONS
CRY303
TRSTCRY303EA 0 NOK
TRSTCRY303
Manual detergent, alkaline, Sapphire
Rengjøringsløsninger Detergenter og overflatevasking
A high foam, heavy duty, detergent for removal of organic soils, including fats, oils and silicone.
  • Alkaline detergent for organic soils
  • Dilution for use: between 1 to 5% v/v
Areas of use include, walls, floors, product contact areas and equipment such as vessels, filling lines and utensils. Sapphire is not soft metal safe, but can be used on a variety of surfaces including stainless steel.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.