Control slide, Fungi TISSUE-TROL™, Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
TTR004-25EA
TTR004-25EAEA 2240 NOK
TTR004-25EA
Control slide, Fungi TISSUE-TROL™, Sigma-Aldrich®
Objektglass Microscopy Control Slides
Histology slides with paraffin embedded section of mouse lung tissue containing Candida albicans.
Suitable for use with Silver Stain (Modified GMS) Procedure No. HT100.
Bestill nå
SPESIFIKASJONER
Arter