TISSUE-TROL™ Control Slides Helicobacter
Leverandør: SIGMA ALDRICH MICROSCOPY
TTR006-25EA
TTR006-25EAEA 2240 NOK
TTR006-25EA
TISSUE-TROL™ Control Slides Helicobacter
Objektglass Microscopy Control Slides
Histology slides with paraffin embedded section of mouse intestine tissue containing Helicobacter pylori. Suitable for use with modified steiner-steiner silver stain procedure No. HT101.
Leveringsinformasjon: Box contains 24 unstained slides plus one reference slide.
Bestill nå
SPESIFIKASJONER
Arter